Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(R)-(+)-Limonen 97%

(R)-(+)-Limonen 97%

Glass bottle, 100 mL
Katalog # L04733.AE
Brandfarlig
Irriterende
Miljø
Leverandør:   Thermo Fisher Scientific
(R)-(+)-Limonen 97%
Katalog # L04733.AE
CAS nummer:  5989-27-5
(R)-(+)-Limonen 97%
(R)-(+)-Limonen 97%

Glass bottle, 100 mL

CAS nummer:  5989-27-5
Price
314,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents

Stabilized

Purity: 97% CAS No.: 5989-27-5 Molecular Formula: C10H16 Formula Weight: 136.23 Flash Point: 48°(118°F) Refractive Index: 1.4730 Odour: Fruit-like Melting Point: -74° Boiling Point: 177-178° Beilstein Reference: 2204754 Merck Reference: 14,5493 Optical Rotation: +115° (c=10 in ethanol) UN No.: 2052 MDL No.: MFCD00062991

Synonymer:  (R)-(+)-1-Methyl-4-(1-methylvinyl)cyclohexen, (R)-(+)-Dipenten, (R)-(+)-p-Mentha-1,8-dien, D(+)-Limonen
  • CAS nummer:5989-27-5
  • Formel:C₁₀H₁₆
  • MDL nummer:MFCD00062991
  • Molecular Weight:136.24 g/mol
  • Merck Index:14,5493
  • Kogende Pt:177…178 °C
  • Smeltende Pt:–74 °C
  • Smeltende Pt:−74 °C
  • Tæthed:0,841
  • Flash Pt:48 °C (118 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:227-813-5
  • FN:2052
  • ADR:3,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    100 mL

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.