Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Eddikesyreanhydrid ≥99%

Eddikesyreanhydrid ≥99%

Glass bottle for solids, 2.500 g
Katalog # L04295.0E
Giftig
Ætsende
Brandfarlig
Leverandør:   Thermo Fisher Scientific
Eddikesyreanhydrid ≥99%
Katalog # L04295.0E
CAS nummer:  108-24-7
Eddikesyreanhydrid ≥99%
Eddikesyreanhydrid ≥99%

Glass bottle for solids, 2.500 g

CAS nummer:  108-24-7
Price
943,00 kr
Log ind for tilgængelighed
1 * 2.500 g
Product Details & Documents

Purity: 99+% CAS No.: 108-24-7 Molecular Formula: (CH3CO)2O Formula Weight: 102.09 Flash Point: 49°(120°F) Refractive Index: 1.3900 Odour: Pungent Melting Point: -73° Boiling Point: 138-140° Beilstein Reference: 385737 Merck Reference: 14,56 UN No.: 1715 MDL No.: MFCD00008705

Synonymer:  Acetyl oxide, Acetyl acetate
  • CAS nummer:108-24-7
  • Formel:(CH₃CO)₂O
  • MDL nummer:MFCD00008705
  • Molecular Weight:102.09 g/mol
  • Merck Index:14,56
  • Kogende Pt:138…140 °C
  • Smeltende Pt:–73 °C
  • Tæthed:1,082
  • Flash Pt:49 °C (120 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-564-8
  • FN:1715
  • ADR:8,II
  • NÅ:01-2119486470-36
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    2.500 g

Specifikationer