Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Eddikesyreanhydrid ≥99%

Eddikesyreanhydrid ≥99%

Glass bottle for solids, 250 g
Katalog # L04295.30
Giftig
Ætsende
Brandfarlig
Leverandør:   Thermo Fisher Scientific
Eddikesyreanhydrid ≥99%
Katalog # L04295.30
CAS nummer:  108-24-7
Eddikesyreanhydrid ≥99%
Eddikesyreanhydrid ≥99%

Glass bottle for solids, 250 g

CAS nummer:  108-24-7
Price
321,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents

Purity: 99+% CAS No.: 108-24-7 Molecular Formula: (CH3CO)2O Formula Weight: 102.09 Flash Point: 49°(120°F) Refractive Index: 1.3900 Odour: Pungent Melting Point: -73° Boiling Point: 138-140° Beilstein Reference: 385737 Merck Reference: 14,56 UN No.: 1715 MDL No.: MFCD00008705

Synonymer:  Acetyl oxide, Acetyl acetate
  • CAS nummer:108-24-7
  • Formel:(CH₃CO)₂O
  • MDL nummer:MFCD00008705
  • Molecular Weight:102.09 g/mol
  • Merck Index:14,56
  • Kogende Pt:138…140 °C
  • Smeltende Pt:–73 °C
  • Tæthed:1,082
  • Flash Pt:49 °C (120 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-564-8
  • FN:1715
  • ADR:8,II
  • NÅ:01-2119486470-36
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.