Orders LinkContactUsLinkComponent
Dækglas, Corning® BioCoat™
Dyrkningsskål, fibronectin-belagt, 22 mm, Glas
  734-1008
   Lav fremstillingspåvirkning
 :   Corning
Dækglas, Corning® BioCoat™
  734-1008
 :  Corning
   Lav fremstillingspåvirkning
Dækglas, Corning® BioCoat™
Dækglas, Corning® BioCoat™
Dyrkningsskål, fibronectin-belagt, 22 mm, Glas
kr2,490.00
1 * 60 Items
These cover glasses have a uniform coating of either poly-L-lysine, poly-D-lysine and laminin, human fibronectin or rat tail collagen.
  • Optically clear
  • Low background fluorescence
  • Non-neurotoxic
Designed for applications where optically clear, thin glass bottoms are needed such as immunofluorescence microscopy, fluorescence imaging of live cells, confocal microscopy, high resolution and inverted microscopy and phase contrast microscopy.
  • Ø:
    22 mm
  • Beskrivelse:
    Dyrkningsskål, fibronectin-belagt
  • Materiale:
    Glas
  • Cat. No.:
    734-1008
  • Miljøcertificering:
    ISO 14001 - Bedford
  • Environmentally Preferable:
    Lav fremstillingspåvirkning