Orders LinkContactUsLinkComponent
Testkits, BOD til Spectroquant®, Supelco®
Testkits, BOD til Spectroquant®, Supelco®
BOD Spectroquant® standard **, 210 mg/l, 10 Tests
Katalog # 1.00718.0001
Leverandør:   Merck
Testkits, BOD til Spectroquant®, Supelco®
Katalog # 1.00718.0001
Leverandør:  Merck
CAS nummer:  
Testkits, BOD til Spectroquant®, Supelco®
Testkits, BOD til Spectroquant®, Supelco®
BOD Spectroquant® standard **, 210 mg/l, 10 Tests
CAS nummer:  
Price
1.280,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*3 BOD næringssaltmikstur  1.00688.0001 påkrævet
Specifikationer
Specifikationer
  • Koncentration:
    210 mg/l
  • Pakn:
    10 Tests
  • Beskrivelse:
    BOD Spectroquant® standard **
  • Cat. No.:
    1.00718.0001
  • M-Clarity quality level:
    MQ100
Specifikationer
Accessories
Produktinformation
  • Katalog #
    Model
    Tilgængelighed
    Price
  • Oxygenreaktionsflaske til 1.14662.0001 (MTC)
    Oxygenreaktionsflaske til 1.14662.0001 (MTC)
    1.14663.0001
    MTC
    Log ind for tilgængelighed
    1 * 1 Items - 782,00 kr