Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Test kits, AOX, Spectroquant®, Supelco®

AOX standard concentrate for dilution to 0,2 - 2,0 mg/L, 20 mg/l, 6 Tests
Katalog # 1.00680.0001
Leverandør:   Merck
Test kits, AOX, Spectroquant®, Supelco®
Katalog # 1.00680.0001
Leverandør:  Merck
CAS nummer:  
Test kits, AOX, Spectroquant®, Supelco®
Test kits, AOX, Spectroquant®, Supelco®

AOX standard concentrate for dilution to 0,2 - 2,0 mg/L, 20 mg/l, 6 Tests

CAS nummer:  
Price
kr871.00
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).

  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times

Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.

*1 AOX Enrichment Set 1.00678.0001 and AOX Sample Preparation Set 1.00677.0001 required.

Specifikationer

Specifikationer

  • Koncentration:
    20 mg/l
  • Pakn:
    6 Tests
  • Beskrivelse:
    AOX standard concentrate for dilution to 0,2 - 2,0 mg/L
  • Cat. No.:
    1.00680.0001
  • M-Clarity quality level:
    MQ100

Specifikationer