Orders LinkContactUsLinkComponent
Test kits, AOX, Spectroquant®, Supelco®
Test kits, AOX, Spectroquant®, Supelco®
AOX prøveforberedelsessæt, -, Iron (III) thiocyanate, Prove, Nova 30/60, Multy, Move, 0,09 L
Catalog # 1.00677.0001
Supplier:   Merck
Test kits, AOX, Spectroquant®, Supelco®
Catalog # 1.00677.0001
Supplier:  Merck
CAS Number:  
Test kits, AOX, Spectroquant®, Supelco®
Test kits, AOX, Spectroquant®, Supelco®
AOX prøveforberedelsessæt, -, Iron (III) thiocyanate, Prove, Nova 30/60, Multy, Move, 0,09 L
CAS Number:  
Price
4.050,00 kr
Log in for availability
1 * 0,09 L
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*1 AOX Enrichment Set 1.00678.0001 and AOX Sample Preparation Set 1.00677.0001 required.
Specifications
Specifications
  • Kemisk metode:
    Iron (III) thiocyanate
  • Pakn:
    0,09 L
  • Beskrivelse:
    AOX prøveforberedelsessæt
  • Til enhed:
    Prove, Nova 30/60, Multy, Move
  • Område:
    -
  • Cat. No.:
    1.00677.0001
  • M-Clarity quality level:
    MQ100