Orders LinkContactUsLinkComponent
Test kits, AOX, Spectroquant®, Supelco®
Test kits, AOX, Spectroquant®, Supelco®
AOX kuvettetest*¹, 0,05 - 2,50 mg/L AOX, Iron (III) thiocyanate, Prove, Nova 30/60, Multy, Move, 25 Tests
Katalog # 1.00675.0001
Leverandør:   Merck
Test kits, AOX, Spectroquant®, Supelco®
Katalog # 1.00675.0001
Leverandør:  Merck
CAS nummer:  
Test kits, AOX, Spectroquant®, Supelco®
Test kits, AOX, Spectroquant®, Supelco®
AOX kuvettetest*¹, 0,05 - 2,50 mg/L AOX, Iron (III) thiocyanate, Prove, Nova 30/60, Multy, Move, 25 Tests
CAS nummer:  
Price
1.990,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*1 AOX Enrichment Set 1.00678.0001 and AOX Sample Preparation Set 1.00677.0001 required.
Specifikationer
Specifikationer
  • Kemisk metode:
    Iron (III) thiocyanate
  • Pakn:
    25 Tests
  • Beskrivelse:
    AOX kuvettetest*¹
  • Til enhed:
    Prove, Nova 30/60, Multy, Move
  • Område:
    0,05 - 2,50 mg/L AOX
  • Cat. No.:
    1.00675.0001
  • M-Clarity quality level:
    MQ100