Test kits, AOX, Spectroquant®, Supelco®
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*1 AOX Enrichment Set 1.00678.0001 and AOX Sample Preparation Set 1.00677.0001 required.
Documents
Certifikater
Søg efter partinummerMerck Segment Brochure
Produktlitteratur
ProductPage_100675
Produktspecifikation
Specifikationer
Specifikationer
- Kemisk metode:Iron (III) thiocyanate
- Pakn:25 Tests
- Beskrivelse:AOX kuvettetest*¹
- Til enhed:Prove, Nova 30/60, Multy, Move
- Område:0,05 - 2,50 mg/L AOX
- Cat. No.:1.00675.0001
- M-Clarity quality level:MQ100