Orders LinkContactUsLinkComponent
Conical separating funnels
Conical separating funnels
1000 ml, 29/32 NS
Katalog # 511-0763
Leverandør:   SciLabware
Conical separating funnels
Katalog # 511-0763
Leverandør:  SciLabware
Conical separating funnels
Conical separating funnels
1000 ml, 29/32 NS
Price
1.410,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass, with glass stopper.
With ground glass stopper and interchangeable PTFE key. Without graduation.
Specifikationer
  • Kappestr.:
    29/32 NS
  • Kapacitet:
    1000 ml
  • Seq:
    05