Buchner funnels
30 mm, 35 ml, P40
Katalog # 511-0802
Leverandør: DIXON SCIENTIFIC
Gå til:
Product Details & Documents
Borosilicate glass
- With sintered glass disc
- Very good chemical resistance
- High temperature resistance
For paper or membrane filtration disc.
Specifikationer
- Plade Ø:30 mm
- Porøsitet:P40
- Kapacitet:35 ml