Draining racks, Azlon®
Set of 11 narrow pegs
Katalog # AZLOWRP002P
Leverandør: SciLabware
Gå til:
Product Details & Documents
Easy to assemble, lightweight polystyrene draining racks designed to be wall mounted.
- Pegs slot into place on the back board and are spaced to allow various sizes of lab ware to fit on to the rack
- Water is collected in the trough at the base of the rack and drains away via a rubber tube which can be directed into a nearby sink or container
Ø/peg: 16 mm
Delivery information: Supplied with wall mounting accessories.
Specifikationer
- Beskrivelse:Set of 11 narrow pegs
- Seq:02