Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumthiosulfat, Supelco®

Natriumthiosulfat, Supelco®

Fibre carton, 50 kg
Katalog # 1.06512.9050
Leverandør:   Merck
Natriumthiosulfat, Supelco®
Katalog # 1.06512.9050
Leverandør:  Merck
CAS nummer:  7772-98-7
Natriumthiosulfat, Supelco®
Natriumthiosulfat, Supelco®

Fibre carton, 50 kg

CAS nummer:  7772-98-7
Price
Spørg for pris
Log ind for tilgængelighed
1 * 50 kg
Product Details & Documents
Synonymer:  Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
  • CAS nummer:7772-98-7
  • Formel:Na₂S₂O₃
  • MDL nummer:MFCD00003499
  • Molecular Weight:158.11 g/mol
  • Merck Index:12,08844
  • Kogende Pt:100 °C (1013 hPa)
  • Smeltende Pt:48 °C
  • Tæthed:1,66 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-867-5
Specifikationer

Specifikationer

  • Pakning:
    Fibre carton
  • Pakn:
    50 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.