Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumhydroxid, Supelco®

Natriumhydroxid, Supelco®

Plastic bottle, 10 kg
Katalog # 1.06467.9010
Ætsende
Leverandør:   Merck
Natriumhydroxid, Supelco®
Katalog # 1.06467.9010
Leverandør:  Merck
CAS nummer:  1310-73-2
Natriumhydroxid, Supelco®

Plastic bottle, 10 kg

CAS nummer:  1310-73-2
Price
kr2,010.00
Log ind for tilgængelighed
1 * 10 kg
Product Details & Documents
Synonymer:  Caustic soda
  • CAS nummer:1310-73-2
  • Formel:NaOH
  • MDL nummer:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Kogende Pt:1390 °C (1013 hPa)
  • Smeltende Pt:323 °C
  • Tæthed:2,13 g/cm³ (20 °C)
  • EINECS:215-185-5
  • ADR:8,II
  •  :M-Clarity™ quality level = MQ200
  • Opbevaringstemperatur:Stue temperatur
  • FN:1823
  • NÅ:01-2119457892-27
Specifikationer

Specifikationstestresultater

  • Solubility:
    1090 g/l (20 °C)
  • Melting Point:
    323 °C
  • Molar Mass:
    40.00 g/mol
  • Boiling Point:
    1390 °C (1013 hPa)
  • Vapor Pressure:
    (20 °C)
  • Density:
    2.13 g/cm3 (20 °C)
  • pH:
    14 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    10 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.