Orders LinkContactUsLinkComponent
n-Hexan, EMPLURA®, Supelco®
n-Hexan, EMPLURA®, Supelco®
Steel barrel, 190 L
Katalog # 1.04368.9190
Brandfarlig
Sundhed
Irriterende
Miljø
Leverandør:   Merck
n-Hexan, EMPLURA®, Supelco®
Katalog # 1.04368.9190
Leverandør:  Merck
CAS nummer:  110-54-3
n-Hexan, EMPLURA®, Supelco®
n-Hexan, EMPLURA®, Supelco®
Steel barrel, 190 L
CAS nummer:  110-54-3
Price
Spørg for pris
Log ind for tilgængelighed
1 * 190 L
Product Details & Documents
EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.
Synonymer:  Hexyl hydride, Hexanen, Hexane
  • CAS nummer:110-54-3
  • Formel:H₃C(CH₂)₄CH₃
  • MDL nummer:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:13,04712
  • Kogende Pt:69 °C (1013 hPa)
  • Smeltende Pt:–94,3 °C
  • Tæthed:0,659 g/cm³ (20 °C)
  • Flash Pt:–22 °C
  • Opbevaringstemperatur:Stue temperatur
  • FN:1208
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ200
  • EINECS:203-777-6
Specifikationer
Specifikationer
  • Pakning:
    Steel barrel
  • Pakn:
    190 L