1,2-Dichlorpropan ≥98%
Glass bottle for solids, 100 g
Purity: 98% CAS No.: 78-87-5 Molecular Formula: C3H6Cl2 Formula Weight: 112.99 Flash Point: 4°(39°F) Refractive Index: 1.4380 Odour: Sweet Melting Point: -100° Boiling Point: 95-97° Beilstein Reference: 1718880 Merck Reference: 14,7854 UN No.: 1279 MDL No.: MFCD00000868
: Propylene dichloride
- :78-87-5
- :ClH₂CCH₂CH₂Cl
- :MFCD00000868
- :112.99 g/mol
- :14,7854
- :95…97 °C
- :–100 °C
- :1,157
- :4 °C (39 °F)
- :Stue temperatur
- :201-152-2
- :1279
- :3,II
- Pakning:Glass bottle for solids
- Pakn:100 g