Orders LinkContactUsLinkComponent
1,2-Dichlorpropan ≥98%
Glass bottle for solids, 100 g
  L03083.22
1,2-Dichlorpropan ≥98%
  L03083.22
 :  78-87-5
1,2-Dichlorpropan ≥98%
1,2-Dichlorpropan ≥98%
Glass bottle for solids, 100 g
 :  78-87-5
773,00 kr
1 * 100 g
Purity: 98% CAS No.: 78-87-5 Molecular Formula: C3H6Cl2 Formula Weight: 112.99 Flash Point: 4°(39°F) Refractive Index: 1.4380 Odour: Sweet Melting Point: -100° Boiling Point: 95-97° Beilstein Reference: 1718880 Merck Reference: 14,7854 UN No.: 1279 MDL No.: MFCD00000868
 :  Propylene dichloride
  •  :78-87-5
  •  :ClH₂CCH₂CH₂Cl
  •  :MFCD00000868
  •  :112.99 g/mol
  •  :14,7854
  •  :95…97 °C
  •  :–100 °C
  •  :1,157
  •  :4 °C (39 °F)
  •  :Stue temperatur
  •  :201-152-2
  •  :1279
  •  :3,II
  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g