Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

VWR®, Dip Slides

Nutrient agar with TTC and malt extract agar, for total count, yeasts and moulds, 100
Katalog # 535104D
Leverandør:   VWR Chemicals
VWR®, Dip Slides
Katalog # 535104D
Leverandør:  VWR Chemicals
CAS nummer:  
VWR®, Dip Slides
VWR®, Dip Slides

Nutrient agar with TTC and malt extract agar, for total count, yeasts and moulds, 100

CAS nummer:  
Price
1.470,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents

Dip slides for microbiological monitoring of surfaces and in liquids. Each slide has media on both sides, with a choice of the same medium, or two different media on the two sides of the slide. This allows sampling of two surfaces with every slide. Testing with dip slides is a safe, reliable, economical and fast indicator of the presence of microorganisms. Dip slides, with a push cap, have a larger contact area.

  • Leveres sterile
  • Det tager kun få sekunder at tage prøven
  • Resultaterne kan nemt aflæses efter blot 24-48 timer
Specifikationer

Specifikationer

  • Pakn:
    100
  • Beskrivelse:
    Nutrient agar with TTC and malt extract agar, for total count, yeasts and moulds
  • Cat. No.:
    535104D
  • Mål:
    30 mm rør, 110 mm høj
  • Kontaktområde:
    10 cm²|10
  • Model:
    Push-on-låg
  • Bakke B×L:
    50×22 mm
  • Bladstørrelse:
    80×28 mm|80×28

Specifikationer