Hand towels, SCOTT®
SCOTT® Performance, single sheet roll, 1 ply, hvid, 200 m × 200 mm, 6 rolls/carton
Katalog # KIMW6665
Leverandør: KIMBERLY CLARK
Gå til:
Product Details & Documents
Disposable hand towels made from AIRFLEX* (a voluminous, textile-like fabric).
- High absorption performance, and tear-proof even when wet
- Single, self-presenting towels for enhanced hygiene and less waste
- Extremely soft
Specifikationer
- Pakket:6 rolls/carton
- Farve:hvid
- Beskrivelse:SCOTT® Performance, single sheet roll, 1 ply
- L×B:200 m × 200 mm
- Seq:04
Accessories
Produktinformation
- Katalog #B×D×HTilgængelighedPrice
- Dispenser for hand towel rolls, AQUARIUS* 6959, white ABS, with windowDispenser for hand towel rolls, AQUARIUS* 6959, white ABS, with window115-1779326×241×430 mmLog ind for tilgængelighed1 * 1 Items - 1.480,00 kr