Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for flat cleanroom mops

Accessories for flat cleanroom mops

SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 18" flat head mops; SSU series handles, 18×4" (45.7×10 cm) mm
Katalog # 129-0538
   Reduktion af affald
Leverandør:   MICRONOVA
Accessories for flat cleanroom mops
Katalog # 129-0538
Leverandør:  MICRONOVA
   Reduktion af affald
Accessories for flat cleanroom mops

SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 18" flat head mops; SSU series handles, 18×4" (45.7×10 cm) mm

Price
3.610,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 18" flat head mops; SSU series handles
  • L×B:
    18×4" (45.7×10 cm) mm
  • Seq:
    23
  • Cat. No.:
    129-0538
  • Environmentally Preferable:
    Reduktion af affald
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.