Orders LinkContactUsLinkComponent
Mops, PowerHead™
Mops, PowerHead™
PowerHead™ absorbent foam,356 mm
Katalog # 129-0523
Leverandør:   MICRONOVA
Mops, PowerHead™
Katalog # 129-0523
Leverandør:  MICRONOVA
Mops, PowerHead™
Mops, PowerHead™
PowerHead™ absorbent foam,356 mm
Price
286,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Mop head is constructed of polyester-covered foam mop heads are available both nonsterile and sterile for critical cleaning. Absorbent foam mop head is for pharmaceutical production and buffer areas. Wide-pore foam mop head and dense urethane foam mop head are for cleanroom support and maintenance areas.
  • Stainless steel PowerHead™ handles hold and wring the various mop heads
  • Autoclavable
  • A quick-disconnect fitting on the handle allows the user to customize length with single-piece and extendable universal handles
Ordering information: All components are ordered separately.
Specifikationer
  • Beskrivelse:
    PowerHead™ absorbent foam,356 mm
  • Seq:
    04