0,00 kr
Ordreindtastning
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
Orders Link
ContactUsLinkComponent
FILTERPAPIR CELLULOSE 1 * 100 Items
FILTERPAPIR CELLULOSE 1 * 100 Items
515-1165
:
Sartorius
FILTERPAPIR CELLULOSE 1 * 100 Items
515-1165
:
Sartorius
FILTERPAPIR CELLULOSE 1 * 100 Items
385,00 kr
1 * 100 Items
-
+
Buffer solutions as per DIN 19266-01: 2000
#: 662-0326
: WTW