Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Phenylboronsyre ≥98%

Phenylboronsyre ≥98%

Glass bottle for solids, 250 g
Katalog # A14257.30
Irriterende
Leverandør:   Thermo Fisher Scientific
Phenylboronsyre ≥98%
Katalog # A14257.30
CAS nummer:  98-80-6
Phenylboronsyre ≥98%
Phenylboronsyre ≥98%

Glass bottle for solids, 250 g

CAS nummer:  98-80-6
Price
8.700,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents

Purity: 98+% CAS No.: 98-80-6 Molecular Formula: C6H7BO2 Formula Weight: 121.93 Melting Point: 214-219° Beilstein Reference: 970972 Merck Reference: 14,1068 MDL No.: MFCD00002103

Synonymer:  Phenylmetaboric acid, Dihydroxy(phenyl)borane, Phenyl boric acid, Benzeneboronic acid
  • CAS nummer:98-80-6
  • Formel:C₆H₇BO₂
  • MDL nummer:MFCD00002103
  • Molecular Weight:121.93 g/mol
  • Merck Index:14,1068
  • Smeltende Pt:214…219 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:202-701-9
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.