pH indicators with integral comparison strips, Whatman™
pH-strip med integreret sammenligning, 1,8 - 3,8
WHAT2626-990
Can be used for the rapid determination of pH values has led to many applications in laboratories and industry.
- Instant pH readings
- Accurate for a wide range of routine pH testing
- Inexpensive
- Convenient and portable for field use
Each test strip has a central segment of indicator dye and, printed alongside, eight or more different colour segments marked with corresponding pH values for matching purposes. The pH test value can be read off by direct comparison of the test strip colour and the colour bars. Excellent for coloured solutions, when any changes in colour of the paper stock are automatically cancelled out.
- pH-område:1,8 - 3,8
- Beskrivelse:pH-strip med integreret sammenligning