Test tubes, with rim, medium wall
150 mm, 18 mm, 1,2 mm
Katalog # 212-0461
Leverandør: MURRAY S
Gå til:
Product Details & Documents
Borosilicate test tubes are suitable for all usual laboratory applications. They will withstand temperature change up to and including boiling of samples. They are also highly chemically resistant.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Specifikationer
- Udv.Ø:18 mm
- Vægtykkelse:1,2 mm
- Længde:150 mm
- Seq:07