Orders LinkContactUsLinkComponent
Test tubes with rim, medium wall
Test tubes with rim, medium wall
100 mm, 12 mm
Katalog # 212-0446
Leverandør:   SciLabware
Test tubes with rim, medium wall
Katalog # 212-0446
Leverandør:  SciLabware
Test tubes with rim, medium wall
Test tubes with rim, medium wall
100 mm, 12 mm
Price
788,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
PYREX®, borosilicate glass.
  • Superior performance in comparison to neutral borosilicate and soda-lime glass
  • Excellent resistance to chemicals and high temperature
High quality test tubes designed and constructed for optimum performance.
Certifications: ISO 4142
Specifikationer
  • Udv.Ø:
    12 mm
  • Længde:
    100 mm
  • Seq:
    03