Orders LinkContactUsLinkComponent
Test tubes with rim, light wall
75 mm, 10 mm, 0,6 mm
  212-7078
 :   MURRAY S
Test tubes with rim, light wall
  212-7078
 :  MURRAY S
Test tubes with rim, light wall
Test tubes with rim, light wall
75 mm, 10 mm, 0,6 mm
304,00 kr
1 * 100 Items
Soda-lime glass test tubes are suitable for general mixing and simple laboratory work, and will withstand moderate warming (water bath).
 : They should not be placed directly into naked flames.
  • Udv.Ø:
    10 mm
  • Vægtykkelse:
    0,6 mm
  • Længde:
    75 mm