Orders LinkContactUsLinkComponent
Sponge swabs, Polywipe®
Sponge swabs, Polywipe®
Polywipe, 110 ml beholder
Katalog # 720-0144
Leverandør:   MEDICAL WIRE EQUIPMENT
Sponge swabs, Polywipe®
Katalog # 720-0144
Leverandør:  MEDICAL WIRE EQUIPMENT
Sponge swabs, Polywipe®
Sponge swabs, Polywipe®
Polywipe, 110 ml beholder
Price
1.460,00 kr
Log ind for tilgængelighed
1 * 30 Items
Product Details & Documents
Non-inhibitory sponge material allows effective sampling of larger areas for environmental and hygiene monitoring. Available dry or pre-moistened with sterile diluent. Polywipes are supplied in peel pouches for immediate use, or in a 110 ml blue-capped container for transfer to the laboratory.
  • Blue colour for visibility
  • Triple wrapped and radiation sterilised
Leveringsinformation: Supplied with individual sterile glove for handling.
Specifikationer
  • Beskrivelse:
    Polywipe, 110 ml beholder