Orders LinkContactUsLinkComponent
Sampling swabs, Dryswab™
Dry swabs, standard tip, Wood/cotton
  720-0127
Sampling swabs, Dryswab™
  720-0127
Sampling swabs, Dryswab™
Sampling swabs, Dryswab™
Dry swabs, standard tip, Wood/cotton
2.570,00 kr
1 * 1.250 Items
These dry swabs are ideal for collecting specimens or samples for analysis or for transferring materials during a diagnostic procedure.
  • Sterile, ready-to-use
  • Available with different shaft and head materials
  • Rayon swabs are particularly suitable for the recovery of microorganisms
Labelled tubes: Each swab comes self-contained and ready to use with its own tube for easy transport to the laboratory. The swab is mounted in the tube plug which serves as a convenient holder so there is no need for direct handling of the swab. Each tube and cap assembly is sealed with a tamper evident label for assurance of sterility and which includes the batch details for traceability. The caps are colour coded according to the type of swab.
Peel pouches: Individual or multiple swabs are packed sterile and ready to use within an easy-to-open tamper evident peel pouch. These are especially suitable where the specimen is to be tested immediately or transferred to another transport container perhaps as part of a test kit. Each peel pouch has product and batch details printed for reference and traceability.
Bulk packed: Swabs are packed loose in autoclavable paper bags of 100.
 : *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
  • Beskrivelse:
    Dry swabs, standard tip
  • Seq:
    08
  • Skaft/hoved:
    Wood/cotton