Bufferløsninger i poser, perpHect®-poser, Orion™
4,01, lyserød, Sachet, 10×15 ml, 10
Leverandør: Thermo Orion
Bufferløsninger i poser, perpHect®-poser, Orion™
Katalog # 662-1143
Leverandør: Thermo Orion
CAS nummer:
Gå til:
Product Details & Documents
Solutions in sachets ensure optimum precision. The sachet provides a perfect liquid seal and good protection from light.
- Disposable
- Convenient, colour coded sachet and solution for ease of use
- No danger of contamination
- Traceable to NIST standards
Colour coded: pH 4,01 buffer is pink, pH 7,00 buffer is yellow, and pH 10,01 buffer is blue.
There is no wasted buffer or secondary reservoir. Each sachet contains lot code, expiration date, and buffer temperature table.
The solutions should only be used once to avoid any danger of contamination. They also satisfy the requirements of the GLP and ISO standards.
Documents
Specifikationer
Specifikationer
- Pakning:Sachet
- Pakn:10
- Farve:lyserød
- Pk Info:10×15 ml
- pH-værdi:4,01
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



