Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Basement membranmatrix, Corning® Matrigel®

Basement membranmatrix, Corning® Matrigel®

Corning® Matrigel® Basement Membrane Matrix, 5 mL
Katalog # 734-0270
   Lav fremstillingspåvirkning
Leverandør:   Corning
Basement membranmatrix, Corning® Matrigel®
Katalog # 734-0270
Leverandør:  Corning
   Lav fremstillingspåvirkning
CAS nummer:  
Basement membranmatrix, Corning® Matrigel®
Basement membranmatrix, Corning® Matrigel®

Corning® Matrigel® Basement Membrane Matrix, 5 mL

CAS nummer:  
Price
2.750,00 kr
Log ind for tilgængelighed
1 * 5 mL
Product Details & Documents

Growth Factor Reduced (GFR) formulation is well suited for applications that require a more highly defined basement membrane preparation.

  • Mouse colonies routinely screened for pathogens via Mouse Antibody Production (MAP) testing
  • Extensive PCR testing is performed to screen for a number of pathogens
  • Tested and found negative for bacteria, fungi, and mycoplasma
  • Protein concentrations determined by Lowry method
  • Endotoxin units measured by Limulus Amoebocyte Lysate assay
  • Matrigel Matrix gel stability tested for a period of 14 days at 37°C
  • Biological activity determined for each lot using a neurite outgrowth assay.

Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

Application
• Cell Growth and Differentiation
• Metabolism/Toxicology Studies
• Invasion Assays
• In Vitro and In Vivo Angiogenesis Assays
• In Vivo Angiogenesis Studies and Augmentation of Tumors in Immunosuppressed Mice

Preparation and Storage
Store at -20°C. Avoid multiple freeze-thaws

Note: For research use only. Not intended for diagnostic or therapeutic procedures.

When warmed, the solution polymerizes to produce a biologically active matrix material resembling the mammalian cellular basement membrane. It is ideal for use with in vitro cell cultures of a broad range of cell types, including both primary and stem cell applications.

Specifikationer

Specifikationer

  • Pakn:
    5 mL
  • Beskrivelse:
    Corning® Matrigel® Basement Membrane Matrix
  • Cat. No.:
    734-0270
  • Miljøcertificering:
    ISO 14001 - Bedford
  • Environmentally Preferable:
    Lav fremstillingspåvirkning

Specifikationer

Complementary Products

Produktinformation

  • Katalog #
    Beskrivelse
    Opløsning
    Model
    Anbefalet arbejdsvolumen
    Membran
    Porestørrelse
    Membran Ø
    Dyrkningsområde
    Pakket
    Ant. brønde
    Tilgængelighed
    Price
  • MLDVMICRO4
    High content imaging system
    25 - 100 nm
    ImageXpress Micro 4
    Log ind for tilgængelighed
    1 * 1 Items - 0,00 kr

  • 735-0364
    High-content micro-confocal imaging system
    25 nm
    MICROCONFOCAL
    Log ind for tilgængelighed
    1 * 1 Items - 0,00 kr

  • MLDVNANOMX
    Imaging system with MetaXpress software
    Image Xpress Nano
    001
    Log ind for tilgængelighed
    1 * 1 Items - 0,00 kr

  • 734-2715
    6-well insert plate
    2,6 ml
    Translucent
    24 mm
    4,67 cm²
    6/plate, 4 plates/case
    01
    Log ind for tilgængelighed
    1 * 24 Items - 1.540,00 kr

  • 734-2763
    Half area microplate, with lid, tissue culture treated
    96
    +
    01
    200 µl
    Log ind for tilgængelighed
    1 * 16 Items - 3.250,00 kr

  • 392-0608
    MicroWell™ plate, flat bottom, Nunclon™ Sphera™ treated surface
    96
    +
    06
    400 µl
    Log ind for tilgængelighed
    1 * 8 Items - 1.500,00 kr

  • 738-0204
    Round bottom, spheroid with lid, ultra-low attachment surface
    96
    +
    01
    300 µl
    Log ind for tilgængelighed
    1 * 5 Items - 1.770,00 kr

  • 734-2967
    Standard dish, with 1× surface treated scaffold insert (32,0×1,6 mm)
    35 mm
    43 cm²
    01
    Log ind for tilgængelighed
    1 * 40 Items - 4.000,00 kr

  • 734-2971
    Standard 12-well plate, 6× surface treated scaffold inserts (21,0×1,6 mm)
    113 cm²
    Individually wrapped
    Log ind for tilgængelighed
    1 * 8 Items - 2.150,00 kr