Basement membranmatrix, Corning® Matrigel®
Growth Factor Reduced (GFR) formulation is well suited for applications that require a more highly defined basement membrane preparation.
- Mouse colonies routinely screened for pathogens via Mouse Antibody Production (MAP) testing
- Extensive PCR testing is performed to screen for a number of pathogens
- Tested and found negative for bacteria, fungi, and mycoplasma
- Protein concentrations determined by Lowry method
- Endotoxin units measured by Limulus Amoebocyte Lysate assay
- Matrigel Matrix gel stability tested for a period of 14 days at 37°C
- Biological activity determined for each lot using a neurite outgrowth assay.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Application
• Cell Growth and Differentiation
• Metabolism/Toxicology Studies
• Invasion Assays
• In Vitro and In Vivo Angiogenesis Assays
• In Vivo Angiogenesis Studies and Augmentation of Tumors in Immunosuppressed Mice
Preparation and Storage
Store at -20°C. Avoid multiple freeze-thaws
Note: For research use only. Not intended for diagnostic or therapeutic procedures.
When warmed, the solution polymerizes to produce a biologically active matrix material resembling the mammalian cellular basement membrane. It is ideal for use with in vitro cell cultures of a broad range of cell types, including both primary and stem cell applications.
Cellular Analysis of 3D Spheroid-Based Tumor Invasion Assays Corning® Matrigel® Basement Membrane, Matrix for 3D Culture In Vitro Corning® Matrigel® Matrix, Frequently Asked Questions Culture and Assay Systems Used for 3D Cell Culture Methods for Implantation of Corning® Matrigel® Matrix into Mice and Tissue Fixation
- Pakn:5 mL
- Beskrivelse:Corning® Matrigel® Basement Membrane Matrix, growth factor reduced (GFR)
- Cat. No.:734-0268
- Miljøcertificering:ISO 14001 - Bedford
- Environmentally Preferable:Lav fremstillingspåvirkning
Complementary Products
- BeskrivelseOpløsningModelAnbefalet arbejdsvolumenMembranPorestørrelseMembran ØDyrkningsområdePakketAnt. brønde
- 734-27156-well insert plate2,6 mlTranslucent24 mm4,67 cm²6/plate, 4 plates/case011 * 24 Items - 1.540,00 kr
- 734-2763Half area microplate, with lid, tissue culture treated96+01200 µl1 * 16 Items - 3.250,00 kr
- 392-0608MicroWell™ plate, flat bottom, Nunclon™ Sphera™ treated surface96+06400 µl1 * 8 Items - 1.500,00 kr
- 738-0204Round bottom, spheroid with lid, ultra-low attachment surface96+01300 µl1 * 5 Items - 1.770,00 kr
- 734-2967Standard dish, with 1× surface treated scaffold insert (32,0×1,6 mm)35 mm43 cm²011 * 40 Items - 4.000,00 kr
- 734-2971Standard 12-well plate, 6× surface treated scaffold inserts (21,0×1,6 mm)113 cm²Individually wrapped1 * 8 Items - 2.150,00 kr



