Charts
C482, –20...+50 °C; 0 - 100 % RH, 31 days
Catalog # 620-0421
Supplier: DICKSON
Jump to:
Product Details & Documents
Documents
DICKC652_TDS_EN
Datasheet
Specifications
- Beskrivelse:C482
- Seq:14
- Område:–20...+50 °C; 0 - 100 % RH
- Optagelsestid:31 days