Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
8-Quinolinol ≥99%

8-Quinolinol ≥99%

Glass bottle for solids, 500 g
Katalog # A14720.36
Sundhed
Irriterende
Leverandør:   Thermo Fisher Scientific
8-Quinolinol ≥99%
Katalog # A14720.36
CAS nummer:  148-24-3
8-Quinolinol ≥99%
8-Quinolinol ≥99%

Glass bottle for solids, 500 g

CAS nummer:  148-24-3
Price
1.860,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 148-24-3 Molecular Formula: C9H7NO Formula Weight: 145.16 Melting Point: 72-76° Boiling Point: 267° Solubility: Almost insoluble in water and diethyl ether. Highly soluble in ethanol, acetone, chloroform, and benzene. Beilstein Reference: 114512 Merck Reference: 14,4843 MDL No.: MFCD00006807

Synonymer:  8-Hydroxyquinoline
  • CAS nummer:148-24-3
  • Formel:C₉H₇NO
  • MDL nummer:MFCD00006807
  • Molecular Weight:145.16 g/mol
  • Merck Index:14,4843
  • Kogende Pt:267 °C
  • Smeltende Pt:72…76 °C
  • Tæthed:1,03 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:205-711-1
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer