Orders LinkContactUsLinkComponent
Stirrer blades, PTFE, Quickfit®
Stirrer blades, PTFE, Quickfit®
Stirrer blade, with 24/29 socket, to fit ST6/7, 76 mm
Katalog # 441-0360
Leverandør:   SciLabware
Stirrer blades, PTFE, Quickfit®
Katalog # 441-0360
Leverandør:  SciLabware
Stirrer blades, PTFE, Quickfit®
Stirrer blades, PTFE, Quickfit®
Stirrer blade, with 24/29 socket, to fit ST6/7, 76 mm
Price
549,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
Stirrer blades made of PTFE, and they can be pivoted on the glass stirrer shaft to allow passage through small apertures.
  • Easily detachable for cleaning and replacement purposes
  • Resistance to chemical attack
Suitable for sweeping close to the vessel walls as they will not cause scratching.
Specifikationer
  • Model:
    Stirrer blade, with 24/29 socket, to fit ST6/7
  • Bredde:
    76 mm
  • Seq:
    02