Orders LinkContactUsLinkComponent
Filtration funnels, Quickfit®
Filtration funnels, Quickfit®
P40, 19/26 NS, 20 mm
Katalog # 201-8684
Leverandør:   SciLabware
Filtration funnels, Quickfit®
Katalog # 201-8684
Leverandør:  SciLabware
Filtration funnels, Quickfit®
Filtration funnels, Quickfit®
P40, 19/26 NS, 20 mm
Price
999,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With sintered glass disc.
Cone connection.
Specifications
  • Konusstr.:
    19/26 NS
  • Plade Ø:
    20 mm
  • Porøsitet:
    P40