Powder funnels, Quickfit®
24/29 NS, 105 mm
201-8296
PYREX®, borosilicate glass, stem with cone.
- Meget god kemisk resistens
- Høj temperaturresistens
Designed for transfer of powders into flasks and vessels.
- Konusstr.:24/29 NS
- Tragt udv. Ø:105 mm
- Seq:03