Soxhlet extractors, Quickfit®
Top 40/38, konus 24/29, rørfilterstørrelse: 33 x 100 mm, 100 ml
201-8378
PYREX®, borosilicate glass.
For extracting soluble components or impurities from solids via repeated soaking/siphoning with solvent.
: Conforms to BS 2071
- Ekstraktionskap.:100 ml
- Beskrivelse:Top 40/38, konus 24/29, rørfilterstørrelse: 33 x 100 mm
- Seq:07



