Soxhlet extractors, Quickfit®
Top 34/35, konus 34/35, rørfilterstørrelse: 28 × 120 mm, 100 ml
Katalog # 201-8377
Leverandør: SciLabware
Gå til:
Product Details & Documents
PYREX®, borosilicate glass.
For extracting soluble components or impurities from solids via repeated soaking/siphoning with solvent.
: Conforms to BS 2071
Specifikationer
- Ekstraktionskap.:100 ml
- Beskrivelse:Top 34/35, konus 34/35, rørfilterstørrelse: 28 × 120 mm
- Seq:05



