Soxhlet extractors, Quickfit®
Top 34/35, konus 24/29, rørfilterstørrelse: 28 × 80 mm, 60 ml
Katalog # 201-8375
Leverandør: SciLabware
Gå til:
Product Details & Documents
PYREX®, borosilicate glass.
For extracting soluble components or impurities from solids via repeated soaking/siphoning with solvent.
: Conforms to BS 2071
Specifikationer
- Ekstraktionskap.:60 ml
- Beskrivelse:Top 34/35, konus 24/29, rørfilterstørrelse: 28 × 80 mm
- Seq:04



