Soxhlet extractors, Quickfit®
Top 29/32, konus 24/29, rørfilterstørrelse: 22 × 80 mm, 40 ml
Katalog # 201-8374
Leverandør: SciLabware
Gå til:
Product Details & Documents
PYREX®, borosilicate glass.
For extracting soluble components or impurities from solids via repeated soaking/siphoning with solvent.
Certifications: Conforms to BS 2071
Specifikationer
- Ekstraktionskap.:40 ml
- Beskrivelse:Top 29/32, konus 24/29, rørfilterstørrelse: 22 × 80 mm
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



