Test tubes with rim, light wall
125 mm, 18 mm, 0,6 mm
Katalog # 212-0556
Leverandør: MURRAY S
Gå til:
Product Details & Documents
Soda-lime glass test tubes are suitable for general mixing and simple laboratory work, and will withstand moderate warming (water bath).
: They should not be placed directly into naked flames.
Specifikationer
- Udv.Ø:18 mm
- Vægtykkelse:0,6 mm
- Længde:125 mm