Orders LinkContactUsLinkComponent
Multiple adapters, Quickfit®
24/29 NS, 24/29 NS
Katalog # 201-8574
Leverandør:   SciLabware
Multiple adapters, Quickfit®
Katalog # 201-8574
Leverandør:  SciLabware
Multiple adapters, Quickfit®
Multiple adapters, Quickfit®
24/29 NS, 24/29 NS
Price
899,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass, straight.
  • Both sockets are the same size
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters with two parallel necks/sockets and a cone.
Specifikationer
  • Konusstr.:
    24/29 NS
  • Kappestr.:
    24/29 NS
  • Seq:
    04