Pipette tips
MLA-type, 10 - 200 µl, +, Transparent, Bulk, 1.000 stk./pose
LPIT151140
- CE-IVD:+
- Pakket:Bulk, 1.000 stk./pose
- Volumen:10 - 200 µl
- Spidstype:MLA-type
- Spits farge:Transparent
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
#: 20864.320
: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83640.320
: VWR Chemicals
Natriumhydroxid 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
#: 31770.294
: VWR Chemicals
Vand, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83645.320
: VWR Chemicals