Orders LinkContactUsLinkComponent
Disposable cuvettes
Disposable cuvettes
Disposable cuvettes, 2,4 – 4,5 ml, 10 mm, Plast
Katalog # JENW83070-30
Leverandør:   Cole-Parmer
Disposable cuvettes
Katalog # JENW83070-30
Leverandør:  Cole-Parmer
Disposable cuvettes
Disposable cuvettes
Disposable cuvettes, 2,4 – 4,5 ml, 10 mm, Plast
Price
3.040,00 kr
Log ind for tilgængelighed
1 * 500 Items
Product Details & Documents
Plastic disposable cuvettes for liquid samples in spectrophotometry.
Specifikationer
  • Længde på lysvej:
    10 mm
  • Volumen:
    2,4 – 4,5 ml
  • Vinduesmateriale:
    Plast
  • Beskrivelse:
    Disposable cuvettes
  • Seq:
    05