Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Sample vials, trident vials, squat or tall

Sample vials, trident vials, squat or tall

Squat, 14,00 ml, 25×48 mm, Yes
Katalog # 212-7068
Leverandør:   MURRAY S
Sample vials, trident vials, squat or tall
Katalog # 212-7068
Leverandør:  MURRAY S
Sample vials, trident vials, squat or tall
Sample vials, trident vials, squat or tall

Squat, 14,00 ml, 25×48 mm, Yes

Price
1.250,00 kr
Log ind for tilgængelighed
1 * 162 Items
Product Details & Documents

Vials made from neutral glass with PE plug stoppers. Suitable for a wide variety of uses in the laboratory, production, more.

  • Robust and economical, will withstand moderate temperature change
  • Wide opening for easy filling and emptying
  • Plug stopper for secure closure

Emballering: For production related reasons, the number of vials per box may vary.

Specifikationer
  • Kasket inkluderet:
    Yes
  • Diameter×H:
    25×48 mm
  • Kapacitet:
    14,00 ml
  • Beskrivelse:
    Squat
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.