Inoculating needles
Nåle, grøn, 20 stk./pose
Leverandør: MEDICAL WIRE EQUIPMENT
Gå til:
Product Details & Documents
PS, sterile. Disposable plastic inoculating needles.
- Moderately flexible
- Colour coded plastic
Documents
Specifikationer
- Pakket:20 stk./pose
- Farve:grøn
- Beskrivelse:Nåle