Orders LinkContactUsLinkComponent
Single channel pipettes, electronic, variable volume, Ovation®
100 - 1000 µl, 1 µl, ±3 - ±0,8 %, <0,6 - <0,15 %, lilla, -
  613-4926
Single channel pipettes, electronic, variable volume, Ovation®
  613-4926
Single channel pipettes, electronic, variable volume, Ovation®
Single channel pipettes, electronic, variable volume, Ovation®
100 - 1000 µl, 1 µl, ±3 - ±0,8 %, <0,6 - <0,15 %, lilla, -
4.520,00 kr
1 * 1 Items
Manually operated pipette with quick set digital volume adjustment.
  • Easy to program and calibrate for different environmental conditions and liquids
  • Pipettes stand flat on the bench so no pipette stands are required
  • Motor-driven volume adjustment for ease of change
  • One-way disposable nozzle with built-in filter to prevent pipette contamination
  • Colour-coded for easy volume recognition
* Certified DNase/RNase free
 : Supplied with a lithium ion battery (giving approximately six months use).
  • Farvekode:
    lilla
  • Pakket:
    -
  • Volumen:
    100 - 1000 µl
  • Forøgelse:
    1 µl
  • Nøjagtighed:
    ±3 - ±0,8 %
  • Usikkerhed:
    <0,6 - <0,15 %
Accessories
  •  
    Beskrivelse
  • 613-4946
    Replacement nozzle for ESC model (0,5 - 20 µl); QS and M models (1 - 10/2 - 20 µl); F1 and F2 models (3 - 20 µl)
    1 * 10 Items - 660,00 kr
  •  
    Beskrivelse
  • 613-4949
    Ekstra batteri
    1 * 1 Items - 154,00 kr
  •  
    Beskrivelse
  • VIST9057-1001
    Seal kit, incl. lubricant, allen wrench, seals and instructions, for Ovation 0,2 - 2 µl (QS), Ovation 1 and 2 µl (F1 and F2)
    1 * 1 Each - 218,00 kr
  •  
    Beskrivelse
  • VIST9057-1002
    Seal kit, incl. lubricant, allen wrench, seals and instructions, for Ovation 1 - 10 µl (QS), Ovation 3 - 10 µl (F1 and F2)
    1 * 1 Each - 218,00 kr
  •  
    Beskrivelse
  • VIST9057-2035
    VITON seal kit for Ovation 20 - 200 µl (QS), 101 - 200 µl (F1 and F2)
    1 * 1 Each - 282,00 kr
  •  
    Beskrivelse
  • VIST9057-3036
    VITON seal kit for Ovation 100 - 1000 µl (QS), 201 - 1000 µl (F1 and F2)
    1 * 1 Each - 282,00 kr
  •  
    Beskrivelse
  • VIST9057-2004
    Seal kit, incl. lubricant, allen wrench, seals and instructions, for Ovation 10 - 100 µl (QS), Ovation 21 - 100 µl (F1 and F2)
    1 * 1 Each - 218,00 kr