Single channel pipettes, electronic, variable volume, Ovation®
20 - 200 µl, 0,2 µl, ±2,5 - ±0,8 %, <1 - <0,15 %, blå, -
Katalog # 613-4925
Leverandør: VISTALAB TECHNOLOGIES
Single channel pipettes, electronic, variable volume, Ovation®
Katalog # 613-4925
Leverandør: VISTALAB TECHNOLOGIES
Gå til:
Product Details & Documents
Manually operated pipette with quick set digital volume adjustment.
- Easy to program and calibrate for different environmental conditions and liquids
- Pipettes stand flat on the bench so no pipette stands are required
- Motor-driven volume adjustment for ease of change
- One-way disposable nozzle with built-in filter to prevent pipette contamination
- Colour-coded for easy volume recognition
* Certified DNase/RNase free
Ordering information: Supplied with a lithium ion battery (giving approximately six months use).
Specifikationer
- Farvekode:blå
- Pakket:-
- Volumen:20 - 200 µl
- Forøgelse:0,2 µl
- Nøjagtighed:±2,5 - ±0,8 %
- Usikkerhed:<1 - <0,15 %
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Replacement nozzle for ESC model (0,5 - 20 µl); QS and M models (1 - 10/2 - 20 µl); F1 and F2 models (3 - 20 µl)Replacement nozzle for ESC model (0,5 - 20 µl); QS and M models (1 - 10/2 - 20 µl); F1 and F2 models (3 - 20 µl)613-4946Log ind for tilgængelighed1 * 10 Items - 660,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Ekstra batteri
Produktinformation
- Katalog #TilgængelighedPrice
- Seal kit, incl. lubricant, allen wrench, seals and instructions, for Ovation 0,2 - 2 µl (QS), Ovation 1 and 2 µl (F1 and F2)Seal kit, incl. lubricant, allen wrench, seals and instructions, for Ovation 0,2 - 2 µl (QS), Ovation 1 and 2 µl (F1 and F2)VIST9057-1001Log ind for tilgængelighed1 * 1 Each - 218,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Seal kit, incl. lubricant, allen wrench, seals and instructions, for Ovation 1 - 10 µl (QS), Ovation 3 - 10 µl (F1 and F2)Seal kit, incl. lubricant, allen wrench, seals and instructions, for Ovation 1 - 10 µl (QS), Ovation 3 - 10 µl (F1 and F2)VIST9057-1002Log ind for tilgængelighed1 * 1 Each - 218,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- VITON seal kit for Ovation 20 - 200 µl (QS), 101 - 200 µl (F1 and F2)VITON seal kit for Ovation 20 - 200 µl (QS), 101 - 200 µl (F1 and F2)VIST9057-2035Log ind for tilgængelighed1 * 1 Each - 282,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- VITON seal kit for Ovation 100 - 1000 µl (QS), 201 - 1000 µl (F1 and F2)VITON seal kit for Ovation 100 - 1000 µl (QS), 201 - 1000 µl (F1 and F2)VIST9057-3036Log ind for tilgængelighed1 * 1 Each - 282,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Seal kit, incl. lubricant, allen wrench, seals and instructions, for Ovation 10 - 100 µl (QS), Ovation 21 - 100 µl (F1 and F2)Seal kit, incl. lubricant, allen wrench, seals and instructions, for Ovation 10 - 100 µl (QS), Ovation 21 - 100 µl (F1 and F2)VIST9057-2004Log ind for tilgængelighed1 * 1 Each - 218,00 kr
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



