Poly-D-Lysine flasks, Corning® BioCoat®
250 ml, 75 cm², Ventileret, Canted, 5 per case
Leverandør: Corning
Poly-D-Lysine flasks, Corning® BioCoat®
Katalog # 734-0192
Leverandør: Corning
Gå til:
Product Details & Documents
Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.
- Attachment and spreading of a variety of cell types
- Cell differentiation and neurite outgrowth
- Attachment of fastidious transfected cell lines
- Support survival of primary neurons in culture
- Serum-free or reduced-serum culture
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Lukkestørrelse:Ventileret
- Dyrkningsareal:75 cm²
- Pakket:5 per case
- Kapacitet:250 ml
- Hals:Canted
- Cat. No.:734-0192
- Miljøcertificering:RECs, ISO 14001 - Kennebunk
- Environmentally Preferable:Lav fremstillingspåvirkning



