Celledyrkningsinserts, Falcon®
Inserts for 12-well plates, sterile, Transparent PET membrane, 0,4 μm
Supplier: Corning
Jump to:
Product Details & Documents
Track-etched PET membranes have a smooth surface and defined cylindrical pores that traverse the membrane.
- Low protein binding PET membrane
- Sterilized by gamma irradiation
- A wide variety of configurations including 6, 12 and 24 well
- A broad selection of membrane pore sizes, 0,4, 1,0, 3,0 and 8,0 µm diameter
- Packed in individual blister packs, 48 inserts/case
- Non-Tissue Culture-treated insert housings prevent promiscuous growth of cells on the insert walls
- Innovative hanging design facilitates pipetting and allows for co-culture
Specifications
- Membran:Transparent PET membrane
- Porestørrelse:0,4 μm
- Beskrivelse:Inserts for 12-well plates, sterile
- Seq:02