Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Celledyrkningsinserts, Falcon®

Inserts for 6-well plates, sterile, Transparent PET membrane, 1,0 μm
Katalog # 734-0040
Leverandør:   Corning
Celledyrkningsinserts, Falcon®
Katalog # 734-0040
Leverandør:  Corning
Celledyrkningsinserts, Falcon®
Celledyrkningsinserts, Falcon®

Inserts for 6-well plates, sterile, Transparent PET membrane, 1,0 μm

Price
2.890,00 kr
Log ind for tilgængelighed
1 * 48 Items
Product Details & Documents

Track-etched PET membranes have a smooth surface and defined cylindrical pores that traverse the membrane.

  • Low protein binding PET membrane
  • Sterilized by gamma irradiation
  • A wide variety of configurations including 6, 12 and 24 well
  • A broad selection of membrane pore sizes, 0,4, 1,0, 3,0 and 8,0 µm diameter
  • Packed in individual blister packs, 48 inserts/case
  • Non-Tissue Culture-treated insert housings prevent promiscuous growth of cells on the insert walls
  • Innovative hanging design facilitates pipetting and allows for co-culture
Specifikationer
  • Membran:
    Transparent PET membrane
  • Porestørrelse:
    1,0 μm
  • Beskrivelse:
    Inserts for 6-well plates, sterile
  • Seq:
    07